Merck
  • (4Z,15Z)-Octadecadienoic Acid Inhibits Glycogen Synthase Kinase-3β and Glucose Production in H4IIE Cells.
(4Z,15Z)-Octadecadienoic Acid Inhibits Glycogen Synthase Kinase-3β and Glucose Production in H4IIE Cells.
Lipids (2017-02-15)
Jun Yoshida, Shota Uesugi, Tetsuaki Kawamura, Ken-Ichi Kimura, Dawei Hu, Shuang Xia, Naoki Toyooka, Masao Ohnishi, Hideki Kawashima
ABSTRACT
Many uncommon non-methylene-interrupted fatty acids (NMI FA) are present in limpet gonads, but their biological properties remain unknown. To investigate new biological effects of naturally occurring NMI FA in eukaryotic cells, the biological activities of structurally analogous (4Z,15Z)-octadecadienoic acid (1), (9Z,20Z)-tricosadienoic acid (2), and (12Z,23Z)-hexacosadienoic acid (3) were examined by using a yeast-based drug-screening system using the Ca
Sigma-Aldrich
TDZD-8, ≥98% (HPLC), needles