- Clinical significance of
Clinical significance of
Oncology letters (2017-08-10)
Sho Nambara, Takaaki Masuda, Taro Tobo, Shinya Kidogami, Hisateru Komatsu, Keishi Sugimachi, Hiroshi Saeki, Eiji Oki, Yoshihiko Maehara, Koshi Mimori
PMID28789412
ABSTRACT
The present authors previously identified a novel candidate tumor suppressor gene, zinc finger protein 750 (
Sigma-Aldrich
Anti-ZNF750 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2
Pricing and availability is not currently available.