Merck
  • High performance separation of quaternary amines using microchip non-aqueous electrophoresis coupled with contactless conductivity detection.
High performance separation of quaternary amines using microchip non-aqueous electrophoresis coupled with contactless conductivity detection.
Journal of chromatography. A (2017-04-12)
Roger Cardoso Moreira, Marilia Sousa Lopes, Iris Medeiros Junior, Wendell K T Coltro
ABSTRACT
This study describes the development of an analytical methodology for the separation of quaternary amines using non-aqueous microchip electrophoresis (NAME) coupled with capacitively coupled contactless conductivity detection (C
Sigma-Aldrich
Tetradodecylammonium bromide, ≥99.0% (AT)