Merck
  • Mouse prolyl oligopeptidase plays a role in trophoblast stem cell differentiation into trophoblast giant cell and spongiotrophoblast.
Mouse prolyl oligopeptidase plays a role in trophoblast stem cell differentiation into trophoblast giant cell and spongiotrophoblast.
Placenta (2017-05-11)
Yuki Maruyama, Shin Matsubara, Atsushi P Kimura
ABSTRACT
Prolyl oligopeptidase (prolyl endopeptidase, Prep), a multifunctional protease hydrolyzing -Pro-X- peptide bonds, is highly expressed in the mouse placenta, but the function during development is not known. We explored the possibility of Prep's involvement in placental differentiation. We cultured trophoblast stem cells (TSCs) derived from the E6.5 mouse embryo and investigated the detailed expression pattern of Prep during their differentiation. Prep-specific inhibitors were added to the TSC culture, and the effect on the differentiation was assessed by microscopic observation and the expression of marker gene for each placental cell. During TSC differentiation for 6 days, Prep was constantly detected at mRNA, protein, and activity levels, and the protein was found mainly in the cytoplasm. The addition of 30 μM and 10 μM SUAM-14746, a Prep-specific inhibitor, effectively inhibited the differentiation into spongiotrophoblasts (SpTs) and trophoblast giant cells (TGCs), while the TSC viability was not affected. 5 μM SUAM-14746 impaired the differentiation into SpTs, and 1 μM SUAM-14746 exhibited no effects. Another Prep-specific inhibitor, KYP-2047, did not affect the differentiation. We confirmed efficient inhibition of Prep enzymatic activity in TSCs by both inhibitors. The dose-dependent effect of SUAM-14746 on TSCs suggests that Prep plays an important role in the differentiation into SpTs and TGCs in the mouse placenta.
Sigma-Aldrich
KYP-2047, ≥95% (HPLC)
Pricing and availability is not currently available.