Merck
  • Structural and serological studies of the O-antigen of the bacterium Proteus mirabilis OXK (serogroup O3) used in the Weil-Felix test.
Structural and serological studies of the O-antigen of the bacterium Proteus mirabilis OXK (serogroup O3) used in the Weil-Felix test.
Biochemistry. Biokhimiia (1997-01-01)
A S Swierzko, M Cedzynski, Knirel YuA, S N Senchenkova, N A Kocharova, A S Shashkov, K I Amano, K Kyohno, W Kaca
ABSTRACT
Sugar analysis and 1H- and 13C-NMR spectroscopic studies showed that various strains of Proteus mirabilis OXK used as antigens in the Weil-Felix test for serodiagnosis of rickettsiosis (scrub typhus) produce lipopolysaccharides (LPSs) with the same O-specific polysaccharide chain having the following structure: [formula: see text] where GlcA and GalA are glucuronic and galacturonic acids, respectively. This polysaccharide which defines the O3 specificity of Proteus and has been found earlier in an unclassified P. mirabilis strain S1959, contains an amide of D-galacturonic acid with L-lysine which plays an important role in manifesting the immunospecificity. A cross-reaction was observed in ELISA between sera from patients with scrub typhus, caused by the bacterium Orientia (Rickettsia) tsutsugamushi, and purified LPS of P. mirabilis OXK, thus suggesting that the common epitope involved in the Weil-Felix test is located on P. mirabilis OXK LPS. Rabbit anti-P. mirabilis OXK antibodies did not cross-react with LPS-lacking O. tsutsugamushi strain Gilliam in dot-blotting and Western blotting, and the nature of the rickettsial antigen responsible for the Weil-Felix reaction remains unknown.
Sigma-Aldrich
Lipopolysaccharides from Proteus mirabilis, purified by phenol extraction
Pricing and availability is not currently available.