Merck
  • Identification of a Novel Liver X Receptor Agonist that Regulates the Expression of Key Cholesterol Homeostasis Genes with Distinct Pharmacological Characteristics.
Identification of a Novel Liver X Receptor Agonist that Regulates the Expression of Key Cholesterol Homeostasis Genes with Distinct Pharmacological Characteristics.
Molecular pharmacology (2017-01-15)
Ni Li, Xiao Wang, Yanni Xu, Yuan Lin, Ningyu Zhu, Peng Liu, Duo Lu, Shuyi Si
ABSTRACT
Activation of liver X receptor (LXR) is associated with cholesterol metabolism and anti-inflammatory processes, which makes it beneficial to antiatherosclerosis therapy. Nevertheless, existing agonists that target LXR, for example TO901317, are related to unwanted side effects. In the present study, using a screening method we identified IMB-808, which displayed potent dual LXR
Sigma-Aldrich
IMB-808, ≥98% (HPLC)
Pricing and availability is not currently available.