- Ammonothermal Synthesis of Novel Nitrides: Case Study on CaGaSiN
Ammonothermal Synthesis of Novel Nitrides: Case Study on CaGaSiN
Chemistry (Weinheim an der Bergstrasse, Germany) (2016-12-04)
Jonas Häusler, Lukas Neudert, Mathias Mallmann, Robin Niklaus, Anna-Carina L Kimmel, Nicolas S A Alt, Eberhard Schlücker, Oliver Oeckler, Wolfgang Schnick
PMID27914179
ABSTRACT
The first gallium-containing nitridosilicate CaGaSiN
MATERIALS