- EZH2 or HDAC1 Inhibition Reverses Multiple Myeloma-Induced Epigenetic Suppression of Osteoblast Differentiation.
EZH2 or HDAC1 Inhibition Reverses Multiple Myeloma-Induced Epigenetic Suppression of Osteoblast Differentiation.
Molecular cancer research : MCR (2017-01-26)
Juraj Adamik, Shunqian Jin, Quanhong Sun, Peng Zhang, Kurt R Weiss, Judith L Anderson, Rebecca Silbermann, G David Roodman, Deborah L Galson
PMID28119431
ABSTRACT
In multiple myeloma, osteolytic lesions rarely heal because of persistent suppressed osteoblast differentiation resulting in a high fracture risk. Herein, chromatin immunoprecipitation analyses reveal that multiple myeloma cells induce repressive epigenetic histone changes at the