Merck
  • The interaction of heme with plakortin and a synthetic endoperoxide analogue: new insights into the heme-activated antimalarial mechanism.
The interaction of heme with plakortin and a synthetic endoperoxide analogue: new insights into the heme-activated antimalarial mechanism.
Scientific reports (2017-04-07)
Marco Persico, Roberto Fattorusso, Orazio Taglialatela-Scafati, Giuseppina Chianese, Ivan de Paola, Laura Zaccaro, Francesca Rondinelli, Marco Lombardo, Arianna Quintavalla, Claudio Trombini, Ernesto Fattorusso, Caterina Fattorusso, Biancamaria Farina
ABSTRACT
In the present work we performed a combined experimental and computational study on the interaction of the natural antimalarial endoperoxide plakortin and its synthetic analogue 4a with heme. Obtained results indicate that the studied compounds produce reactive carbon radical species after being reductively activated by heme. In particular, similarly to artemisinin, the formation of radicals prone to inter-molecular reactions should represent the key event responsible for Plasmodium death. To our knowledge this is the first experimental investigation on the reductive activation of simple antimalarial endoperoxides (1,2-dioxanes) by heme and results were compared to the ones previously obtained from the reaction with FeCl
Sigma-Aldrich
2,3-Dimethylhydroquinone, 97%