Merck
  • Multicomponent Droplet Evaporation on Chemical Micro-Patterned Surfaces.
Multicomponent Droplet Evaporation on Chemical Micro-Patterned Surfaces.
Scientific reports (2017-02-06)
Minghao He, Dong Liao, Huihe Qiu
ABSTRACT
The evaporation and dynamics of a multicomponent droplet on a heated chemical patterned surface were presented. Comparing to the evaporation process of a multicomponent droplet on a homogenous surface, it is found that the chemical patterned surface can not only enhance evaporation by elongating the contact line, but also change the evaporation process from three regimes for the homogenous surface including constant contact line (CCL) regime, constant contact angle (CCA) regime and mix mode (MM) to two regimes, i.e. constant contact line (CCL) and moving contact line (MCL) regimes. The mechanism of contact line stepwise movement in MCL regimes in the microscopic range is investigated in detail. In addition, an improved local force model on the contact line was employed for analyzing the critical receding contact angles on homogenous and patterned surfaces. The analysis results agree well for both surfaces, and confirm that the transition from CCL to MCL regimes indicated droplet composition changes from multicomponent to monocomponent, providing an important metric to predict and control the dynamic behavior and composition of a multicomponent droplet using a patterned surface.
Sigma-Aldrich
1H,1H,2H,2H-Perfluorodecyltriethoxysilane, 97%