Merck
  • Interaction of alpha1-adrenoceptor subtypes with different G proteins induces opposite effects on cardiac L-type Ca2+ channel.
Interaction of alpha1-adrenoceptor subtypes with different G proteins induces opposite effects on cardiac L-type Ca2+ channel.
Circulation research (2008-05-10)
Jin O-Uchi, Hiroyuki Sasaki, Satoshi Morimoto, Yoichiro Kusakari, Hitomi Shinji, Toru Obata, Kenichi Hongo, Kimiaki Komukai, Satoshi Kurihara
ABSTRACT
We examined the effect of alpha(1)-adrenoceptor subtype-specific stimulation on L-type Ca2+ current (I(Ca)) and elucidated the subtype-specific intracellular mechanisms for the regulation of L-type Ca2+ channels in isolated rat ventricular myocytes. We confirmed the protein expression of alpha(1A)- and alpha(1B)-adrenoceptor subtypes at the transverse tubules (T-tubules) and found that simultaneous stimulation of these 2 receptor subtypes by nonsubtype selective agonist, phenylephrine, showed 2 opposite effects on I(Ca) (transient decrease followed by sustained increase). However, selective alpha(1A)-adrenoceptor stimulation (> or =0.1 micromol/L A61603) only potentiated I(Ca), and selective alpha(1B)-adrenoceptor stimulation (10 mumol/L phenylephrine with 2 micromol/L WB4101) only decreased I(Ca). The positive effect by alpha(1A)-adrenoceptor stimulation was blocked by the inhibition of phospholipase C (PLC), protein kinase C (PKC), or Ca2+/calmodulin-dependent protein kinase II (CaMKII). The negative effect by alpha(1B)-adrenoceptor stimulation disappeared after the treatment of pertussis toxin or by the prepulse depolarization, but was not attributable to the inhibition of cAMP-dependent pathway. The translocation of PKCdelta and epsilon to the T-tubules was observed only after alpha(1A)-adrenoceptor stimulation, but not after alpha(1B)-adrenoceptor stimulation. Immunoprecipitation analysis revealed that alpha(1A)-adrenoceptor was associated with G(q/11), but alpha(1B)-adrenoceptor interacted with one of the pertussis toxin-sensitive G proteins, G(o). These findings demonstrated that the interactions of alpha(1)-adrenoceptor subtypes with different G proteins elicit the formation of separate signaling cascades, which produce the opposite effects on I(Ca). The coupling of alpha(1A)-adrenoceptor with G(q/11)-PLC-PKC-CaMKII pathway potentiates I(Ca). In contrast, alpha(1B)-adrenoceptor interacts with G(o), of which the betagamma-complex might directly inhibit the channel activity at T-tubules.
Product Description
Supelco
WB-4101 hydrochloride, analytical standard
Pricing and availability is not currently available.