Merck
  • Switchable Synthesis of Pyrroles and Pyrazines via Rh(II)-Catalyzed Reaction of 1,2,3-Triazoles with Isoxazoles: Experimental and DFT Evidence for the 1,4-Diazahexatriene Intermediate.
Switchable Synthesis of Pyrroles and Pyrazines via Rh(II)-Catalyzed Reaction of 1,2,3-Triazoles with Isoxazoles: Experimental and DFT Evidence for the 1,4-Diazahexatriene Intermediate.
The Journal of organic chemistry (2016-12-16)
Nikolai V Rostovskii, Julia O Ruvinskaya, Mikhail S Novikov, Alexander F Khlebnikov, Ilia A Smetanin, Anastasiya V Agafonova
ABSTRACT
4-Aminopyrrole-3-carboxylates and pyrazine-2-carboxylates were synthesized from 5-alkoxyisoxazoles and 1-sulfonyl-1,2,3-triazoles by tuning the Rh(II) catalyst and the reaction conditions. The reaction in chloroform at 100 °C under Rh
Sigma-Aldrich
Isoxazole, 99%