Merck
  • Targeting autophagy augments the activity of DHA-E3 to overcome p-gp mediated multi-drug resistance.
Targeting autophagy augments the activity of DHA-E3 to overcome p-gp mediated multi-drug resistance.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2016-11-09)
Guangmin Xi, Ming Wang, Bing Sun, Abdul Sami Shaikh, Yongqing Liu, Wei Wang, Hongxiang Lou, Huiqing Yuan
ABSTRACT
Multidrug resistance (MDR) is a major obstacle for successful chemotherapy treatment. Searching for effective MDR modulators and combining them with anticancer drug therapies has been a promising strategy against clinical MDR. In our previous study, we have found that DHA-E3, a synthetic derivative of DHA, has the ability to modulate the function of P-glycoprotein (P-gp) and reverse MDR in cancer cells. In this study, we further evaluated the reversal effect of DHA-E3 on MDR and explored its mechanism of action in vitro. Our findings showed that DHA-E3 significantly potentiated the cytotoxicity of vincristine(VCR) and adriamycin(ADR) in the P-gp over-expressing KB/VCR and A02 cells. The mechanistic study found that DHA-E3 increased the intracellular accumulation of ADR and rhodamine-123 by directly inhibiting the drug-transport activity of P-gp. In the present study, we found that DHA-E3 not only reversed MDR, but also induced autophagy in MDR cancer cells. To determine whether DHA-E3-induced autophagy is an adaptive survival response or contributes to cell death, we manipulated autophagic activity using autophagy inhibitor 3-MA or siRNA targeting Beclin1. We found that the reversal activity of DHA-E3 was significantly exacerbated in the presence of 3-MA or blocking the expression of Beclin1. These results suggest that DHA-E3 is capable of reversing MDR, induction of autophagy represents a defense mechanism and inhibiting this process may be an effective strategy to augment the reversal activity of reversal agents.
Sigma-Aldrich
MISSION® esiRNA, targeting human BECN1
Pricing and availability is not currently available.