- Nanosized molecular propellers by cyclodehydrogenation of polyphenylene dendrimers.
Nanosized molecular propellers by cyclodehydrogenation of polyphenylene dendrimers.
Journal of the American Chemical Society (2004-03-12)
Christopher D Simpson, Gunter Mattersteig, Kai Martin, Lileta Gherghel, Roland E Bauer, Hans Joachim Räder, Klaus Müllen
PMID15012144
ABSTRACT
In a polymer analogous approach, large dendritic oligophenylenes containing benzene and tetraphenylmethane cores are transformed via oxidative cyclodehydrogenation to novel propeller-shaped molecules with large polycyclic aromatic hydrocarbon units as "blades". Structure analysis is performed by a combination of MALDI-TOF mass spectrometry, UV/vis, fluorescence, and Raman spectroscopy using solid-state sample preparation methods. These methods are also utilized to determine the degree of the cyclodehydrogenation reaction.