- Synthesis and anti-HIV activity of 2-naphthyl substituted DAPY analogues as non-nucleoside reverse transcriptase inhibitors.
Synthesis and anti-HIV activity of 2-naphthyl substituted DAPY analogues as non-nucleoside reverse transcriptase inhibitors.
Nine newly 6-cyano-2-naphthyl substituted diarylpyrimidines (DAPY) were synthesized as non-nucleoside reverse transcriptase inhibitors on the basis of our previous work. The antiviral and cytotoxicity evaluation indicated that these compounds displayed strong activity against wild-type HIV-1 at nanomolar concentrations with selectivity index SI greater than 23 779. The most active compounds 3c and 3e exhibited activity against the double mutant (103N+181C) strains at an EC₅₀ of 0.16 and 0.15 μM, and were more activity than that of efavirenz.