Merck
  • Improvement and biological applications of fluorescent probes for zinc, ZnAFs.
Improvement and biological applications of fluorescent probes for zinc, ZnAFs.
Journal of the American Chemical Society (2002-06-06)
Tomoya Hirano, Kazuya Kikuchi, Yasuteru Urano, Tetsuo Nagano
ABSTRACT
The development and cellular applications of novel fluorescent probes for Zn2+, ZnAF-1F, and ZnAF-2F are described. Fluorescein is used as a fluorophore of ZnAFs, because its excitation and emission wavelengths are in the visible range, which minimizes cell damage and autofluorescence by excitation light. N,N-Bis(2-pyridylmethyl)ethylenediamine, used as an acceptor for Zn2+, is attached directly to the benzoic acid moiety of fluorescein, resulting in very low quantum yields of 0.004 for ZnAF-1F and 0.006 for ZnAF-2F under physiological conditions (pH 7.4) due to the photoinduced electron-transfer mechanism. Upon the addition of Zn2+, the fluorescence intensity is quickly increased up to 69-fold for ZnAF-1F and 60-fold for ZnAF-2F. Apparent dissociation constants (K(d)) are in the nanomolar range, which affords sufficient sensitivity for biological applications. ZnAFs do not fluoresce in the presence of other biologically important cations such as Ca2+ and Mg2+, and are insensitive to change of pH. The complexes with Zn2+ of previously developed ZnAFs, ZnAF-1, and ZnAF-2 decrease in fluorescence intensity below pH 7.0 owing to protonation of the phenolic hydroxyl group of fluorescein, whose pKa value is 6.2. On the other hand, the Zn2+ complexes of ZnAF-1F and ZnAF-2F emit stable fluorescence around neutral and slightly acidic conditions because the pKa values are shifted to 4.9 by substitution of electron-withdrawing fluorine at the ortho position of the phenolic hydroxyl group. For application to living cells, the diacetyl derivative of ZnAF-2F, ZnAF-2F DA, was synthesized. ZnAF-2F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-2F, which is retained in the cells. Using ZnAF-2F DA, we could measure the changes of intracellular Zn2+ in cultured cells and hippocampal slices.
Sigma-Aldrich
ZnAF-2F, ≥95.0% (HPCE)