Merck
  • Anticancer effect of eupatilin on glioma cells through inhibition of the Notch-1 signaling pathway.
Anticancer effect of eupatilin on glioma cells through inhibition of the Notch-1 signaling pathway.
Molecular medicine reports (2015-12-18)
Yawei Wang, Hongwei Hou, Ming Li, Yang Yang, Lan Sun
ABSTRACT
Eupatilin, one of the major flavonoids in Artemisia asiatica Nakai (Asteraceae), has been reported to possess antitumor properties. However, thus far there have been no reports regarding the effects of eupatilin on glioma. Therefore, in the current study the effects of eupatilin on glioma and the underlying molecular mechanism were explored. The effect of eupatilin on cell viability was detected by the MTT assay. Cell invasion and migration were performed with Transwell assays and cell apoptosis was determined by flow cytometric analysis. Notch‑1 knockdown cells were established by transfection with Notch‑1 small interfering RNA (siRNA). The expression levels of Notch‑1 were detected by quantitative reverse transcription‑polymerase chain reaction and western blotting. The results of the present study indicated that eupatilin exhibits an anticancer effect on glioma cells. Eupatilin inhibited proliferation, reduced cell invasion and migration, and promoted the apoptosis of glioma cells. Additionally, it suppressed Notch‑1 expression. Knockdown of Notch‑1 by siRNA contributed to the inhibitory effect of eupatilin on proliferation and invasion of glioma cells. In conclusion, eupatilin had an inhibitory effect on proliferation, invasion and migration, and promoted apoptosis of glioma cells through suppression of the Notch‑1 signaling pathway. Therefore, eupatilin may have potential as an effective agent for the treatment of glioma.
Sigma-Aldrich
Eupatilin, ≥98% (HPLC)
Pricing and availability is not currently available.