Merck
  • Effect of cytokines on Siglec-1 and HIV-1 entry in monocyte-derived macrophages: the importance of HIV-1 envelope V1V2 region.
Effect of cytokines on Siglec-1 and HIV-1 entry in monocyte-derived macrophages: the importance of HIV-1 envelope V1V2 region.
Journal of leukocyte biology (2015-12-17)
Ousman Jobe, Hung V Trinh, Jiae Kim, Wadad Alsalmi, Sodsai Tovanabutra, Philip K Ehrenberg, Kristina K Peachman, Guofen Gao, Rasmi Thomas, Jerome H Kim, Nelson L Michael, Carl R Alving, Venigalla B Rao, Mangala Rao
ABSTRACT
Monocytes and monocyte-derived macrophages express relatively low levels of CD4. Despite this, macrophages can be effectively infected with human immunodeficiency virus type 1. Macrophages have a critical role in human immunodeficiency virus type 1 transmission; however, the mechanism or mechanisms of virus infection are poorly understood. We report that growth factors, such as granulocyte macrophage colony-stimulating factor and macrophage colony-stimulating factor affect the phenotypic profile and permissiveness of macrophages to human immunodeficiency virus type 1. Human immunodeficiency virus type 1 infection of monocyte-derived macrophages derived from granulocyte macrophage and macrophage colony-stimulating factors was predominantly facilitated by the sialic acid-binding immunoglobulin-like lectin-1. The number of sialic acid-binding immunoglobulin-like lectin receptors on macrophage colony-stimulating factor-derived monocyte-derived macrophages was significantly greater than on granulocyte macrophage colony-stimulating factor-derived monocyte-derived macrophages, and correspondingly, human immunodeficiency virus type 1 infection was greater in the macrophage colony-stimulating factor-derived monocyte-derived macrophages. Single-genome analysis and quantitative reverse transcriptase-polymerase chain reaction revealed that the differences in infectivity was not due to differences in viral fitness or in viral variants with differential infectivity but was due to reduced viral entry into the granulocyte macrophage colony-stimulating factor-derived monocyte-derived macrophages. Anti-sialic acid-binding immunoglobulin-like lectin, trimeric glycoprotein 145, and scaffolded V1V2 proteins were bound to sialic acid-binding immunoglobulin-like lectin and significantly reduced human immunodeficiency virus type 1 entry and infection. Furthermore, sialic acid residues present in the V1V2 region of the envelope protein mediated human immunodeficiency virus type 1 interaction with sialic acid-binding immunoglobulin-like lectin and entry into macrophage colony-stimulating factor-derived monocyte-derived macrophages. Removal of sialic acid residues or glycans from scaffolded V1V2 protein decreased human immunodeficiency virus type 1 infectivity. These results highlight the importance of sialic acids on the V1V2 region in binding to sialic acid-binding immunoglobulin-like lectin and suggest that the unusually long surface-exposed sialic acid-binding immunoglobulin-like lectin might aid in the capture and entry of human immunodeficiency virus type 1 into monocyte-derived macrophages.
Sigma-Aldrich
6′-Sialyllactose sodium salt, from bovine milk or colostrum, ≥97% (HPLC)