Merck
  • Implementation of meiosis prophase I programme requires a conserved retinoid-independent stabilizer of meiotic transcripts.
Implementation of meiosis prophase I programme requires a conserved retinoid-independent stabilizer of meiotic transcripts.
Nature communications (2016-01-09)
Emilie Abby, Sophie Tourpin, Jonathan Ribeiro, Katrin Daniel, Sébastien Messiaen, Delphine Moison, Justine Guerquin, Jean-Charles Gaillard, Jean Armengaud, Francina Langa, Attila Toth, Emmanuelle Martini, Gabriel Livera
ABSTRACT
Sexual reproduction is crucially dependent on meiosis, a conserved, specialized cell division programme that is essential for the production of haploid gametes. Here we demonstrate that fertility and the implementation of the meiotic programme require a previously uncharacterized meiosis-specific protein, MEIOC. Meioc invalidation in mice induces early and pleiotropic meiotic defects in males and females. MEIOC prevents meiotic transcript degradation and interacts with an RNA helicase that binds numerous meiotic mRNAs. Our results indicate that proper engagement into meiosis necessitates the specific stabilization of meiotic transcripts, a previously little-appreciated feature in mammals. Remarkably, the upregulation of MEIOC at the onset of meiosis does not require retinoic acid and STRA8 signalling. Thus, we propose that the complete induction of the meiotic programme requires both retinoic acid-dependent and -independent mechanisms. The latter process involving post-transcriptional regulation likely represents an ancestral mechanism, given that MEIOC homologues are conserved throughout multicellular animals.
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Pricing and availability is not currently available.