Merck
  • IkappaB kinase-2-independent and -dependent inflammation in airway disease models: relevance of IKK-2 inhibition to the clinic.
IkappaB kinase-2-independent and -dependent inflammation in airway disease models: relevance of IKK-2 inhibition to the clinic.
Molecular pharmacology (2006-03-07)
Mark A Birrell, Sissie Wong, Elizabeth L Hardaker, Matthew C Catley, Kerryn McCluskie, Michael Collins, Saleem Haj-Yahia, Maria G Belvisi
ABSTRACT
Nuclear factor kappaB (NF-kappaB) is a transcription factor believed to be central in the expression of numerous inflammatory genes and the pathogenesis of many respiratory diseases. We have previously demonstrated increased NF-kappaB pathway activation in a steroid-sensitive animal model of lipopolysaccharide (LPS)-driven airway inflammation. It is noteworthy that this phenomenon was not observed in a steroid-insensitive model of elastase-induced inflammation in the rat. The aim of this study was to gather further evidence to suggest that these similar profiles of neutrophilic inflammation can be NF-kappaB-dependent or -independent by determining the impact of an IkappaB kinase-2 (IKK-2) inhibitor, 2-[(aminocarbonyl)amino]-5-(4-fluorophenyl)-3-thiophenecarboxamide (TPCA-1). In the LPS model, TPCA-1 blocked the increase in NF-kappaB DNA binding, a marker of NF-kappaB pathway activation. This inhibition was associated with a reduction in inflammatory mediator release [tumor necrosis factor alpha (TNFalpha)/interleukin-1beta (IL-1beta)/matrix metalloproteinase-9 (MMP-9)] and lung inflammatory cell burden (neutrophilia/eosinophilia). These data were paralleled with a steroid and in human cell based assays. In the elastase-driven inflammation model, in which our group has previously failed to measure an increase in NF-kappaB DNA binding, neither TPCA-1 nor the steroid, affected mediator release (IL-1beta/MMP-9) or cellular burden (neutrophilia/lymphomononuclear cells). This is the first study to examine the effect of an IKK-2 inhibitor in well validated models that mimic aspects of the inflammatory lesion evident in diseases such as COPD. In conclusion, we have demonstrated that animal models with similar profiles of airway inflammation can be IKK-2 inhibitor/steroid-sensitive or -insensitive. If both profiles of inflammation exist in the clinic, then this finding is extremely exciting and may lead to greater understanding of disease pathology and the discovery of novel anti-inflammatory targets.
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)