Merck
  • 1H fast MAS NMR studies of hydrogen-bonding interactions in self-assembled monolayers.
1H fast MAS NMR studies of hydrogen-bonding interactions in self-assembled monolayers.
Journal of the American Chemical Society (2003-04-03)
Shane Pawsey, Mark McCormick, Susan De Paul, Robert Graf, Y S Lee, Linda Reven, Hans W Spiess
ABSTRACT
The structures formed by the adsorption of carboxyalkylphosphonic acids on metal oxides were investigated by (1)H fast magic angle spinning (MAS), heteronuclear correlation (HETCOR), and (1)H double-quantum (DQ) MAS solid-state NMR experiments. The diacids HO(2)C(CH(2))(n)PO(3)H(2) (n = 2, 3, 11, and 15) were adsorbed on TiO(2) and two types of ZrO(2) powders having average particle sizes of 20, 30, and 5 nm, respectively. Carboxyalkylphosphonic acids bind selectively via the phosphonate group, forming monolayers with pendant carboxylic acid groups. Whereas dipolar coupled P-OH protons are detected on TiO(2), there are only isolated residual P-OH groups on ZrO(2), reflecting the relative binding strengths of phosphonic acids on these two substrates. From a comparative (1)H MAS NMR study with an analogous monolayer system, HO(2)C(CH(2))(7)SH coated gold nanoparticles, the hydrogen-bonding network at the monolayer/air interface is found to be quite disordered, at least for SAMs deposited on nonplanar substrates. Whereas only hydrogen-bonded homodimers occur in the bulk diacids, hydrogen bonding between the carboxylic and phosphonic acid groups is present in multilayers of the diacids on the ZrO(2) nanopowder.
Sigma-Aldrich
Methyl 16-bromohexadecanoate, 97%