Merck
Marinopyrrole A target elucidation by acyl dye transfer.
Journal of the American Chemical Society (2009-08-14)
Chambers C Hughes, Yu-Liang Yang, Wei-Ting Liu, Pieter C Dorrestein, James J La Clair, William Fenical
ABSTRACT
The targeting of marinopyrrole A to actin was identified using a fluorescent dye transfer strategy. The process began by appending a carboxylic acid terminal tag to a phenol in the natural product. The resulting probe was then studied in live cells to verify that it maintained activity comparable to marinopyrrole A. Two-color fluorescence microscopy confirmed that both unlabeled and labeled materials share comparable uptake and subcellular localization in HCT-116 cells. Subsequent immunoprecipitation studies identified actin as a putative target in HCT-116 cells, a result that was validated by mass spectral, affinity, and activity analyses on purified samples of actin. Further data analyses indicated that the dye in the marinopyrrole probe was selectively transferred to a single residue K(115), an event that did not occur with related acyl phenols and reactive labels. In this study, the combination of cell, protein, and amino acid analysis arose from a single sample of material, thereby, suggesting a means to streamline and reduce material requirements involved in mode of action studies.
Sigma-Aldrich
(±)-Marinopyrrole A, ≥95% (HPLC)