Merck
  • Disruption of the retinoblastoma pathway by small interfering RNA and ectopic expression of the catalytic subunit of telomerase lead to immortalization of human ovarian surface epithelial cells.
Disruption of the retinoblastoma pathway by small interfering RNA and ectopic expression of the catalytic subunit of telomerase lead to immortalization of human ovarian surface epithelial cells.
Oncogene (2006-09-06)
G Yang, D G Rosen, J A Colacino, I Mercado-Uribe, J Liu
ABSTRACT
The risk of developing ovarian cancer is about 1% over a lifetime, but it is the most deadly gynecologic cancer, in part due to lack of diagnostic markers for early-stage disease and cell model system for studying early neoplastic changes. Most existing immortal human ovarian surface epithelial cells were achieved by using viral protein such as SV40 T/t antigen or E6/E7, which inactivate multiple cellular pathways. In the current study, we used a small interfering RNA (siRNA) against the retinoblastoma gene (pRb) and ectopic expression of human telomerase reverse transcriptase (hTERT) to immortalize the primary ovarian epithelial cell line OSE137 and two additional human ovarian surface epithelial cells. The immortalized OSE137 showed increased telomerase activity, lengthened telomeres, increased G2/M phase, altered cell-cycle regulatory proteins but nontumorigenic. As both Rb and hTERT pathways are commonly altered in human ovarian cancer and these genetic changes are faithfully modeled in these cells without using viral protein, these immortal cells represent an authentic in vitro model system with which to study the initiation and progression of human ovarian cancer.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Roche
TeloTAGGG Telomere Length Assay, sufficient for ≤50 reactions, kit of 1 (15 components), suitable for cell culture