Merck
  • Benchtop monitoring of reaction progress via visual recognition with a handheld UV lamp: in situ monitoring of boronic acids in the Suzuki-Miyaura reaction.
Benchtop monitoring of reaction progress via visual recognition with a handheld UV lamp: in situ monitoring of boronic acids in the Suzuki-Miyaura reaction.
Organic letters (2006-12-29)
Timothy E Barder, Stephen L Buchwald
ABSTRACT
[reaction: see text] Although boronic acids are widely used in metal-catalyzed reactions, it is difficult to assay their consumption. As such, we developed a reversible fluorescent sensor that is activated upon binding a boronic acid. The sensor can be used to monitor consumption of a boronic acid in Suzuki-Miyaura reactions. Importantly, only a standard handheld long-wave UV lamp (365 nm) is required and fluorescence is easily detectable with the naked eye without disturbing the reaction mixture.
Sigma-Aldrich
7,8-Dihydroxy-4-methylcoumarin, 97%
Pricing and availability is not currently available.