Merck
  • A single nucleotide in stem loop II of 5'-untranslated region contributes to virulence of enterovirus 71 in mice.
A single nucleotide in stem loop II of 5'-untranslated region contributes to virulence of enterovirus 71 in mice.
PloS one (2011-11-10)
Ming-Te Yeh, Shainn-Wei Wang, Chun-Keung Yu, Kuei-Hsiang Lin, Huan-Yao Lei, Ih-Jen Su, Jen-Ren Wang
ABSTRACT
Enterovirus 71 (EV71) has emerged as a neuroinvasive virus responsible for several large outbreaks in the Asia-Pacific region while virulence determinant remains unexplored. In this report, we investigated increased virulence of unadapted EV71 clinical isolate 237 as compared with isolate 4643 in mice. A fragment 12 nucleotides in length in stem loop (SL) II of 237 5'-untranslated region (UTR) visibly reduced survival time and rate in mice was identified by constructing a series of infectious clones harboring chimeric 5'-UTR. In cells transfected with bicistronic plasmids, and replicon RNAs, the 12-nt fragment of isolate 237 enhanced translational activities and accelerated replication of subgenomic EV71. Finally, single nucleotide change from cytosine to uridine at base 158 in this short fragment of 5'-UTR was proven to reduce viral translation and EV71 virulence in mice. Results collectively indicated a pivotal role of novel virulence determinant C158 on virus translation in vitro and EV71 virulence in vivo. These results presented the first reported virulence determinant in EV71 5'-UTR and first position discovered from unadapted isolates.
Sigma-Aldrich
Anti-Enterovirus 71 Antibody, cross-reacts with Coxsackie A16, clone 422-8D-4C-4D, ascites fluid, clone 422-8D-4C-4D, Chemicon®
Pricing and availability is not currently available.