Merck
  • Effect of paroxetine hydrochloride (Paxil) on fatigue and depression in breast cancer patients receiving chemotherapy.
Effect of paroxetine hydrochloride (Paxil) on fatigue and depression in breast cancer patients receiving chemotherapy.
Breast cancer research and treatment (2005-03-09)
Joseph A Roscoe, Gary R Morrow, Jane T Hickok, Karen M Mustian, Jennifer J Griggs, Sara E Matteson, Peter Bushunow, Raman Qazi, Brian Smith
ABSTRACT
Fatigue can significantly interfere with a cancer patient's ability to fulfill daily responsibilities and enjoy life. It commonly co-exists with depression in patients undergoing chemotherapy, suggesting that administration of an antidepressant that alleviates symptoms of depression could also reduce fatigue. We report on a double-blind clinical trial of 94 female breast cancer patients receiving at least four cycles of chemotherapy randomly assigned to receive either 20 mg of the selective serotonin re-uptake inhibitor (SSRI) paroxetine (Paxil, SmithKline Beecham Pharmaceuticals) or an identical-appearing placebo. Patients began their study medication seven days following their first on-study treatment and continued until seven days following their fourth on-study treatment. Seven days after each treatment, participants completed questionnaires measuring fatigue (Multidimensional Assessment of Fatigue, Profile of Mood States-Fatigue/Inertia subscale and Fatigue Symptom Checklist) and depression (Profile of Mood States-Depression subscale [POMS-DD] and Center for Epidemiologic Studies-Depression [CES-D]). Repeated-measures ANOVAs, after controlling for baseline measures, showed that paroxetine was more effective than placebo in reducing depression during chemotherapy as measured by the CES-D (p = 0.006) and the POMS-DD (p = 0.07) but not in reducing fatigue (all measures, ps > 0.27). Although depression was significantly reduced in the 44 patients receiving paroxetine compared to the 50 patients receiving placebo, indicating that a biologically active dose was used, no significant differences between groups on any of the measures of fatigued were observed. Results suggest that modulation of serotonin may not be a primary mechanism of fatigue related to cancer treatment.
Sigma-Aldrich
Paroxetine hydrochloride hemihydrate, ≥98% (HPLC), powder