MilliporeSigma
  • Sensitive detection of norovirus using phage nanoparticle reporters in lateral-flow assay.
Sensitive detection of norovirus using phage nanoparticle reporters in lateral-flow assay.
PloS one (2015-05-16)
Anna E V Hagström, Gavin Garvey, Andrew S Paterson, Sagar Dhamane, Meena Adhikari, Mary K Estes, Ulrich Strych, Katerina Kourentzi, Robert L Atmar, Richard C Willson
ABSTRACT
Noroviruses are recognized worldwide as the principal cause of acute, non-bacterial gastroenteritis, resulting in 19-21 million cases of disease every year in the United States. Noroviruses have a very low infectious dose, a short incubation period, high resistance to traditional disinfection techniques and multiple modes of transmission, making early, point-of-care detection essential for controlling the spread of the disease. The traditional diagnostic tools, electron microscopy, RT-PCR and ELISA require sophisticated and expensive instrumentation, and are considered too laborious and slow to be useful during severe outbreaks. In this paper we describe the development of a new, rapid and sensitive lateral-flow assay using labeled phage particles for the detection of the prototypical norovirus GI.1 (Norwalk), with a limit of detection of 107 virus-like particles per mL, one hundred-fold lower than a conventional gold nanoparticle lateral-flow assay using the same antibody pair.