Merck
  • Cytotoxic and xenoestrogenic effects via biotransformation of trans-anethole on isolated rat hepatocytes and cultured MCF-7 human breast cancer cells.
Cytotoxic and xenoestrogenic effects via biotransformation of trans-anethole on isolated rat hepatocytes and cultured MCF-7 human breast cancer cells.
Biochemical pharmacology (2003-06-24)
Yoshio Nakagawa, Toshinari Suzuki
ABSTRACT
The metabolism and action of trans-anethole (anethole) and the estrogen-like activity of the compound and its metabolites were studied in freshly isolated rat hepatocytes and cultured MCF-7 human breast cancer cells, respectively. The incubation of hepatocytes with anethole (0.25-2.0mM) caused a concentration- and time-dependent cell death accompanied by losses of cellular ATP and adenine nucleotide pools. Anethole at a weakly toxic level (0.5mM) was metabolized to 4-methoxycinnamic acid (4MCA), 4-hydroxy-1-propenylbenzene (4OHPB), and the monosulfate conjugate of 4OHPB; the levels of 4OHPB sulfate and 4MCA reached approximately 20 and 200 microM within 2 hr, respectively, whereas that of free unconjugated 4OHPB was less than approximately 0.5 microM. At a moderately toxic concentration (1.0mM), unconjugated 4OHPB reached approximately 10 microM, followed by abrupt loss of 3'-phosphoadenosine 5'-phosphosulphate (PAPS). Based on cell viability and adenine nucleotide levels, 4OHPB was more toxic than anethole and 4MCA. The addition of 2,6-dichloro-4-nitrophenol (50 microM), an inhibitor of sulfotransferase, enhanced the anethole-induced cytotoxicity associated with losses of ATP, PAPS, and 4OHPB sulfate, and symmetrically increased the unconjugated 4OHPB concentration. 4OHPB as well as diethylstilbestrol (DES) and bisphenol A (BPA), which are known xenoestrogenic compounds, competitively displaced 17beta-estradiol bound to the estrogen receptor alpha in a concentration-dependent manner; IC(50) values of these compounds were approximately 1 x 10(-5), 1 x 10(-8) and 5 x 10(-5)M, respectively. 4OHPB also caused a concentration (10(-8) to 10(-6)M)-dependent proliferation of MCF-7 cells, whereas neither anethole nor 4MCA (10(-9) to 10(-5)M) affected cell proliferation. However, at higher concentrations (>10(-4)M), 4OHPB rather than anethole and 4MCA was cytotoxic. These results suggest that the biotransformation of anethole induces a cytotoxic effect at higher concentrations in rat hepatocytes and an estrogenic effect at lower concentrations in MCF-7 cells based on the concentrations of the hydroxylated intermediate, 4OHPB.
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%