Merck
  • A new synthesis of naphthyridinones and quinolinones: palladium-catalyzed amidation of o-carbonyl-substituted aryl halides.
A new synthesis of naphthyridinones and quinolinones: palladium-catalyzed amidation of o-carbonyl-substituted aryl halides.
Organic letters (2004-07-02)
Peter J Manley, Mark T Bilodeau
ABSTRACT
[reaction: see text] An alternative to the Friedlander condensation for the synthesis of naphthyridinones and quinolinones has been discovered. Palladium-catalyzed amidation of halo aromatics substituted in the ortho position by a carbonyl functional group or its equivalent with primary or secondary amides leads to the formation of substituted naphthyridinones and quinolinones.
Sigma-Aldrich
Xantphos, 97%