Merck
  • Novel terphenyls as selective cyclooxygenase-2 inhibitors and orally active anti-inflammatory agents.
Novel terphenyls as selective cyclooxygenase-2 inhibitors and orally active anti-inflammatory agents.
Journal of medicinal chemistry (1996-04-26)
J J Li, M B Norton, E J Reinhard, G D Anderson, S A Gregory, P C Isakson, C M Koboldt, J L Masferrer, W E Perkins, K Seibert, Y Zhang, B S Zweifel, D B Reitz
ABSTRACT
A novel series of terphenyl methyl sulfones and sulfonamides have been shown to be highly potent and selective cyclooxygenase-2 (COX-2) inhibitors. The sulfonamide analogs 17 and 21 were found to be much more potent COX-2 inhibitors and orally active anti-inflammatory agents than the corresponding methyl sulfone analogs 16 and 20, respectively, albeit with some decrease in COX-2 selectivity. Structure-activity relationship studies have determined that incorporation of two fluorine atoms in the central phenyl group, as in 20 and 21, is extremely advantageous for both in vitro COX-2 potency and selectivity as well as in vivo activity. Several noticeable examples in the 1,2-diaryl-4,5-difluorobenzenesulfonamide series are 21a-c,k,l,n (COX-2, IC50 = 0.002-0.004 microM), in which all have in vitro COX-1/COX-2 selectivity > 1000. In addition, sulfonamides 21a,b,d,g,j,m,n,q were shown to have greatly enhanced oral activity with more than 90% inhibition of prostaglandin E2 production in the air pouch model of inflammation. Furthermore, sulfonamide 21b was found to be very active in the rat adjuvant-induced arthritis model (ED50 = 0.05 mg/kg) and carrageenan-induced hyperalgesia assay (ED50 = 38.7 mg/kg) with no indication of gastrointestinal toxicity in rats at doses as high as 200 mg/kg.
Sigma-Aldrich
4-Fluorophenylboronic acid, ≥95%
Pricing and availability is not currently available.