Merck
  • Phenelfamycins G and H, new elfamycin-type antibiotics produced by Streptomyces albospinus Acta 3619.
Phenelfamycins G and H, new elfamycin-type antibiotics produced by Streptomyces albospinus Acta 3619.
The Journal of antibiotics (2011-02-03)
Elke Brötz, Andreas Kulik, Sabaratnam Vikineswary, Chew-Theng Lim, Geok Yuan Annie Tan, Heidi Zinecker, Johannes F Imhoff, Thomas Paululat, Hans-Peter Fiedler
ABSTRACT
Phenelfamycins G and H are new members of the family of elfamycin antibiotics with the basic structure of phenelfamycins E and F, respectively, which are also well known as ganefromycins α and β. Phenelfamycins G and H differ from phenelfamycins E and F by an additional hydroxy group at position C-30, which is not described so far for any of the elfamycin-type antibiotics. The actinomycete strain that produced phenelfamycins G and H was identified to be Streptomyces albospinus based on its 16S rRNA gene sequence. Phenelfamycins G and H exhibit a narrow antibacterial spectrum with a pronounced inhibitory activity against Propionibacterium acnes.
Product Description
Roche
Reverse Transcriptase Assay, colorimetric, suitable for enzyme immunoassay, kit of 1 (14 components), sufficient for ≤200 tests
Pricing and availability is not currently available.