Merck
  • Liver failure attributable to pyrrolizidine alkaloid toxicosis and associated with inspiratory dyspnea in ponies: three cases (1982-1988).
Liver failure attributable to pyrrolizidine alkaloid toxicosis and associated with inspiratory dyspnea in ponies: three cases (1982-1988).
Journal of the American Veterinary Medical Association (1991-05-01)
E G Pearson
ABSTRACT
Of 41 equids referred to a veterinary teaching hospital in the Pacific northwest because of dyspnea and inspiratory noise, 3 ponies were diagnosed as having liver failure, most likely attributable to pyrrolizidine alkaloid toxicosis. Dyspnea appeared to be caused by laryngeal and/or pharyngeal paralysis. It is proposed that this paralysis was a manifestation of hepatic encephalopathy. Although these clinical signs are not common for pyrrolizidine toxicosis, practitioners should be aware of the possibility so that misdiagnosis of other causes of inspiratory dyspnea may not be made.
Sigma-Aldrich
Iodonitrotetrazolium chloride, 95%
Pricing and availability is not currently available.