Merck
  • Simvastatin decreases steroid production in the H295R cell line and decreases steroids and FSH in female rats.
Simvastatin decreases steroid production in the H295R cell line and decreases steroids and FSH in female rats.
Reproductive toxicology (Elmsford, N.Y.) (2015-10-18)
Anna Guldvang, Cecilie Hurup Hansen, Johan J Weisser, Bent Halling-Sørensen, Bjarne Styrishave
ABSTRACT
Endocrine modulating effects of Simvastatin (SV) and its metabolite, Simvastatin β-hydroxy acid (SVA), were investigated in H295R cells and in female Sprague-Dawley (SPRD) rats. H295R cells were exposed to SV and SVA concentrations from 0 to 10μM for 48h. Four groups of SPRD rats received 0 (CT), 1.3 (L), 5.0 (M), and 20.0 (H)mg SV/kg bw/day for 14 days. 10 Steroids were investigated in H295R growth media, and in tissues and plasma from rats using GC-MS/MS. Plasma LH and FSH were quantified by ELISA. In the H295R assay, SV and SVA particularly decreased progestagens with IC50-values from 0.10-0.13μM for SV and from 0.019-0.055μM for SVA. In rats, SV decreased progestagens in ovaries, brain and plasma, and plasma FSH in the M (72.4% decrease) and H group (76.6% decrease). Because progestagens and gonadotropins are major players in fertility, administration of SV might exert negative effects on female reproduction.
Product Description
Supelco
Tenivastatin ammonium salt, analytical standard
Pricing and availability is not currently available.