Merck
  • BMP-2 induces motility and invasiveness by promoting colon cancer stemness through STAT3 activation.
BMP-2 induces motility and invasiveness by promoting colon cancer stemness through STAT3 activation.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine (2015-07-01)
Bo Ram Kim, Sang Cheul Oh, Dae-Hee Lee, Jung Lim Kim, Suk Young Lee, Myoung Hee Kang, Sun Il Lee, Sanghee Kang, Sung Yup Joung, Byung Wook Min
ABSTRACT
Bone morphogenetic proteins (BMPs) have been involved in metastatic progression and tumorigenesis of many cancer types. However, it remains unclear how BMP-2 contributes to the initiation and development of these cancers. Here, we investigated the role of BMP-2 in colon cancer stem cell (CSC) development from colon cancer cells. We also determined the effects of BMP-2 on CSC development and epithelial-mesenchymal transition (EMT) in human colon cancer cell lines HCT-116 and SW620. We found that BMP-2 enhanced sphere formation of colon cancer cells without serum. Also, BMP-2-induced spheres displayed up-regulation of stemness markers (CD133+ and EpCAM+) and increased drug resistance, hallmarks of CSCs. Importantly, expression of EMT activators p-Smad1/5 and Snail and N-cadherin was increased in the spheres' cells, indicating that BMP-2 signaling might result in CSC self-renewal and EMT. Furthermore, siRNA-mediated knockdown of signal transducer and activator of transcription 3 (STAT3) in HCT-116 cells reversed BMP-2-induced EMT and stem cell formation. Taken together, our results suggest that the BMP-2 induced STAT3-mediated induction of colon cancer cell metastasis requires an EMT and/or changes in CSC markers.
Sigma-Aldrich
MISSION® esiRNA, targeting human SMAD1
Sigma-Aldrich
2-Phenylindole, technical grade, 95%