- New library of aminosulfonyl-tagged Hoveyda-Grubbs type complexes: Synthesis, kinetic studies and activity in olefin metathesis transformations.
New library of aminosulfonyl-tagged Hoveyda-Grubbs type complexes: Synthesis, kinetic studies and activity in olefin metathesis transformations.
Beilstein journal of organic chemistry (2010-12-18)
Etienne Borré, Frederic Caijo, Christophe Crévisy, Marc Mauduit
PMID21165173
ABSTRACT
Seven novel Hoveyda-Grubbs precatalysts bearing an aminosulfonyl function are reported. Kinetic studies indicate an activity enhancement compared to Hoveyda's precatalyst. A selection of these catalysts was investigated with various substrates in ring-closing metathesis of dienes or enynes and cross metathesis. The results demonstrate that these catalysts show a good tolerance to various chemical functions.