Merck
  • Structure-activity relationship of imidazo[1,2-a]pyridines as ligands for detecting beta-amyloid plaques in the brain.
Structure-activity relationship of imidazo[1,2-a]pyridines as ligands for detecting beta-amyloid plaques in the brain.
Journal of medicinal chemistry (2003-01-10)
Zhi-Ping Zhuang, Mei-Ping Kung, Alan Wilson, Chi-Wan Lee, Karl Plössl, Catherine Hou, David M Holtzman, Hank F Kung
ABSTRACT
A series of novel beta-amyloid (A beta) aggregate-specific ligands, 2-(4'-dimethylaminophenyl)-6-iodoimidazo[1,2-a]pyridine, 16(IMPY), and its related derivatives were prepared. An in vitro binding study with preformed A beta aggregates showed that 16(IMPY) and its bromo derivative competed with binding of 2-(4'-dimethylaminophenyl)-6-iodobenzothiazole, [125I]7(TZDM), a known ligand for A beta aggregates, with high binding affinities (K(i) = 15 and 10 nM, respectively). In vitro autoradiography of brain sections of a transgenic mouse (Tg2576) with [125I]16(IMPY) displayed high selective binding to amyloid-like structures, comparable to that observed by staining with thioflavin-S visualized under fluorescence. In vivo biodistribution after an intravenous injection of [125I]16(IMPY) in normal mice showed a high initial brain uptake and fast washout, indicating a low background activity associated with this iodinated ligand. Taken together, the data suggests that [123I]16(IMPY) may be useful for imaging A beta aggregates in patients with Alzheimer's disease.
Sigma-Aldrich
2-Amino-5-iodopyridine, 98%