Merck
Graphene oxide: efficiency of reducing agents.
Chemical communications (Cambridge, England) (2013-07-19)
Siegfried Eigler, Stefan Grimm, Michael Enzelberger-Heim, Paul Müller, Andreas Hirsch
ABSTRACT
The efficiency of reducing agents for the reduction of graphene oxide (GO) could be probed by scanning Raman spectroscopy. A film of graphene flakes derived from GO was probed to be graphene like. We also focus on the surface quality of reduced GO (rGO).
Sigma-Aldrich
Hydriodic acid, contains ≤1.5% hypophosphorous acid as stabilizer, ACS reagent, ≥47.0%