Merck
  • Curcuminoids and acetylacetone: iron complexes as studied by electrospray ionization mass spectrometry.
Curcuminoids and acetylacetone: iron complexes as studied by electrospray ionization mass spectrometry.
European journal of mass spectrometry (Chichester, England) (2015-04-24)
Wojciech Ostrowski
ABSTRACT
Curcumin complexes with iron ions were investigated by electrospray ionization mass spectrometry. It was shown that in methanol solutions of curcumin and iron(III) ions, complexes are formed with a stoichiometry of 1 : 1, 2 : or 3 : 1. Curcumi-iron(II) complexes were created only with the 1 : 1 stoichiometry. Abundances of the 1 : 1 ions depend on the source of iron [(II) or (III)] and the reduction process of iron(III) by higher cone- voltage values. A counter-ion for the iron cation does not affect the types of formed complexes. For comparison, methanol solutions of acetylacetone (acac) and mixtures of acac and curcumin with iron salts were also investigated.
Sigma-Aldrich
Demethoxycurcumin, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Bisdemethoxycurcumin, ≥98% (HPLC), solid
Pricing and availability is not currently available.
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Pricing and availability is not currently available.
Sigma-Aldrich
Curcumin, ≥94% (curcuminoid content), ≥80% (Curcumin)
Pricing and availability is not currently available.