Merck
  • Identification of the critical features of a small peptide inhibitor of JNK activity.
Identification of the critical features of a small peptide inhibitor of JNK activity.
The Journal of biological chemistry (2002-01-16)
Renae K Barr, Tulene S Kendrick, Marie A Bogoyevitch
ABSTRACT
The c-Jun N-terminal kinases (JNKs) are a subfamily of the mitogen-activated protein kinases (MAPKs). Although progress in evaluating the functions of other MAPKs has been facilitated by the characterization of specific inhibitors, no JNK-directed inhibitor is commercially available. We have identified a 21-amino acid peptide inhibitor of activated JNKs, based on amino acids 143-163 of the JNK-binding domain (JBD) of the JNK scaffolding protein, JNK-interacting protein-1 (JIP-1). This peptide, I-JIP (Inhibitor of JNK-based on JIP-1), inhibited JNK activity in vitro toward recombinant c-Jun, Elk, and ATF2 up to 90%. A truncated I-JIP (TI-JIP), the C-terminal 11 amino acids of I-JIP, directly interacted with recombinant JNKs but not its substrates as shown by surface plasmon resonance analysis. Scanning alanine replacement within truncated I-JIP identified 4 residues (Arg-156, Pro-157, Leu-160, or Leu-162) as independently critical for inhibition. JBD peptide sequences from JIP-2 and JIP-3 shared these critical residues and accordingly were effective JNK inhibitors. In contrast, peptides based on the JBDs of ATF2 and c-Jun inhibited JNK activity by <40%, which agreed with their lack of homology to the critical Arg-156 and Pro-157. These studies thus define a small peptide inhibitor sequence of JNKs based on the JIP proteins.
Sigma-Aldrich
JNK Inhibitor I, (L)-Form, Cell-Permeable, The JNK Inhibitor I, (L)-Form, Cell-Permeable controls the biological activity of JNK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.