Merck
  • Influence of Alkoxy Groups on the Photoinduced Dynamics of Organic Molecules Exemplified on Alkyl Vinyl Ethers.
Influence of Alkoxy Groups on the Photoinduced Dynamics of Organic Molecules Exemplified on Alkyl Vinyl Ethers.
The journal of physical chemistry. A (2015-10-23)
O Schalk, M Stenrup, T Geng, R Lindh, R D Thomas, R Feifel, T Hansson
ABSTRACT
A series of different alkyl vinyl ethers is investigated to decipher the possible reaction channels upon photoexcitation to the π3s-Rydberg and the ππ*-valence state at 200 nm using time-resolved photoelectron spectroscopy and on-the-fly time-dependent density functional theory dynamics simulations. The results indicate two possible relaxation pathways: (1) a radiationless decay through the ππ*-state back to the ground state via torsion of the C═C double bond, in accordance with the dynamics found in ethylene; and (2) a fast dissociation of the C-O bond between the alkyl and the vinoxy group in the πσ*-state. The latter state can be accessed only after excitation to the π3s-Rydberg state (quantum yield of ∼50% according to the dynamics simulations). Additionally, the excited state barrier leading to formation of a vinyl radical was found to be too high to be crossed. These results indicate that the dynamics of ethers crucially depend on the excitation wavelength and that the πσ*-state constitutes an important competitive reaction channel that leads to dissociation of the molecules.
Sigma-Aldrich
Propyl vinyl ether, 99%
Sigma-Aldrich
Ethyl vinyl ether, contains 0.1% KOH as stabilizer, 99%