Merck
Bmi-1 expression modulates non-small cell lung cancer progression.
Cancer biology & therapy (2015-04-17)
Dan Xiong, Yunlin Ye, Yujie Fu, Jinglong Wang, Bohua Kuang, Hongbo Wang, Xiumin Wang, Lidong Zu, Gang Xiao, Mingang Hao, Jianhua Wang
ABSTRACT
Previous studies indicate that the role of B lymphoma Mo-MLV insertion region 1 homolog (Bmi-1) is responsible for multiple cancer progression. However, Bmi-1 in controlling gene expression in non-small cell lung cancer (NSCLC) development is not well explored. Here we report that the Bmi-1 level is highly increased in primary NSCLC tissues compared to matched adjacent non-cancerous tissues and required for lung tumor growth in xenograft model. Furthermore, we also demonstrate that Bmi-1 level is lower in matched involved lymph node cancerous tissues than the respective primary NSCLC tissues. We find that Bmi-1 does not affect cell cycle and apoptosis in lung cancer cell lines as it does not affect the expression of p16/p19, Pten, AKT and P-AKT. Mechanistic analyses note that reduction of Bmi-1 expression inversely regulates invasion and metastasis of NSCLC cells in vitro and in vivo, followed by induction of epithelial-mesenchymal transition (EMT). Using genome microarray assays, we find that RNAi-mediated silence of Bmi-1 modulates some important molecular genetics or signaling pathways, potentially associated with NSCLC development. Taken together, our findings disclose for the first time that Bmi-1 level accumulates strongly in early stage and then declines in late stage, which is potentially important for NSCLC cell invasion and metastasis during progression.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Bmi1
Sigma-Aldrich
MISSION® esiRNA, targeting human BMI1, COMMD3-BMI1