Merck
  • Quantitative interaction proteomics and genome-wide profiling of epigenetic histone marks and their readers.
Quantitative interaction proteomics and genome-wide profiling of epigenetic histone marks and their readers.
Cell (2010-09-21)
Michiel Vermeulen, H Christian Eberl, Filomena Matarese, Hendrik Marks, Sergei Denissov, Falk Butter, Kenneth K Lee, Jesper V Olsen, Anthony A Hyman, Henk G Stunnenberg, Matthias Mann
ABSTRACT
Trimethyl-lysine (me3) modifications on histones are the most stable epigenetic marks and they control chromatin-mediated regulation of gene expression. Here, we determine proteins that bind these marks by high-accuracy, quantitative mass spectrometry. These chromatin "readers" are assigned to complexes by interaction proteomics of full-length BAC-GFP-tagged proteins. ChIP-Seq profiling identifies their genomic binding sites, revealing functional properties. Among the main findings, the human SAGA complex binds to H3K4me3 via a double Tudor-domain in the C terminus of Sgf29, and the PWWP domain is identified as a putative H3K36me3 binding motif. The ORC complex, including LRWD1, binds to the three most prominent transcriptional repressive lysine methylation sites. Our data reveal a highly adapted interplay between chromatin marks and their associated protein complexes. Reading specific trimethyl-lysine sites by specialized complexes appears to be a widespread mechanism to mediate gene expression.