Merck
  • An arsenite-inducible 19S regulatory particle-associated protein adapts proteasomes to proteotoxicity.
An arsenite-inducible 19S regulatory particle-associated protein adapts proteasomes to proteotoxicity.
Molecular cell (2006-09-16)
Ariel Stanhill, Cole M Haynes, Yuhong Zhang, Guangwei Min, Matthew C Steele, Juliya Kalinina, Enid Martinez, Cecile M Pickart, Xiang-Peng Kong, David Ron
ABSTRACT
Protein misfolding caused by exposure to arsenite is associated with transcriptional activation of the AIRAP gene. We report here that AIRAP is an arsenite-inducible subunit of the proteasome's 19S cap that binds near PSMD2 at the 19S base. Compared to the wild-type, knockout mouse cells or C. elegans lacking AIRAP accumulate more polyubiquitylated proteins and exhibit higher levels of stress when exposed to arsenite, and proteasomes isolated from arsenite-treated AIRAP knockout cells are relatively impaired in substrate degradation in vitro. AIRAP's association with the 19S cap reverses the stabilizing affect of ATP on the 26S proteasome during particle purification, and AIRAP-containing proteasomes, though constituted of 19S and 20S subunits, acquire features of hybrid proteasomes with both 19S and 11S regulatory caps. These features include enhanced cleavage of peptide substrates and suggest that AIRAP adapts the cell's core protein degradation machinery to counteract proteotoxicity induced by an environmental toxin.
Sigma-Aldrich
Diamide
Pricing and availability is not currently available.