Merck
  • Prolonged administration of antidepressant drugs leads to increased binding of [(3)H]MPEP to mGlu5 receptors.
Prolonged administration of antidepressant drugs leads to increased binding of [(3)H]MPEP to mGlu5 receptors.
Neuropharmacology (2014-05-07)
Gabriel Nowak, Lucyna Pomierny-Chamioło, Agata Siwek, Ewa Niedzielska, Bartosz Pomierny, Agnieszka Pałucha-Poniewiera, Andrzej Pilc
ABSTRACT
Metabotropic glutamate 5 (mGlu5) receptors are functionally connected with NMDA receptors. The antidepressant activity of the NMDA receptor antagonist ketamine in both preclinical and clinical studies, along with the antidepressant-like activities of negative allosteric modulators (NAMs) of mGlu5, led us to investigate if prolonged administration of various antidepressant drugs or the mGlu5 NAM, MTEP, causes changes in mGlu5 receptor availability or protein expression or in expression of Homer proteins in the rat brain. Our results clearly show that prolonged treatment with antidepressants with various mechanisms of action (such as escitalopram, reboxetine, milnacipran, moclobemide and imipramine) or with MTEP led to significant increases in [(3)H]MPEP binding in homogenates of the hippocampus and/or cerebral cortex. Increases in mGlu5 expression were also observed, though they did not always parallel the increase in binding. The results indicate that adaptive up-regulation of mGlu5 receptors may be a common change induced by antidepressant drugs.
Sigma-Aldrich
Milnacipran hydrochloride, ≥98% (HPLC), solid
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
6-Methyl-2-(phenylethynyl)pyridine hydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Escitalopram oxalate, ≥98% (HPLC), powder
USP
Escitalopram oxalate, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)