Merck
  • Cibacron blue immobilized poly(glycidyl-methacrylate) nanobeads for albumin removal in proteome studies.
Cibacron blue immobilized poly(glycidyl-methacrylate) nanobeads for albumin removal in proteome studies.
Artificial cells, nanomedicine, and biotechnology (2013-10-31)
Müge Andaç
ABSTRACT
A new approach to albumin removal for proteome studies involving human plasma samples was presented with dye affinity poly glycidyl methacrylate (PGMA) nanobeads in an average size of 45 nm. The specific surface area of PGMA nanobeads was calculated as 2616 m(2)/g. Cibacron Blue F3GA (CB) was immobilized onto PGMA nanobeads as dye ligand, and CB immobilized PGMA (CB-PGMA) nanobeads were characterized by a serial processing. Albumin depletion efficiency of CB-PGMA nanobeads was investigated in human plasma sample and confirmed by two-dimensional gel electrophoresis.
Sigma-Aldrich
Glycidyl methacrylate, ≥97.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Glycidyl methacrylate, 97%, contains 100 ppm monomethyl ether hydroquinone as inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
Plasma, from bovine
Pricing and availability is not currently available.