Merck
  • Specific inhibition of HDAC4 in cardiac progenitor cells enhances myocardial repairs.
Specific inhibition of HDAC4 in cardiac progenitor cells enhances myocardial repairs.
American journal of physiology. Cell physiology (2014-06-20)
Ling X Zhang, Megan DeNicola, Xin Qin, Jianfeng Du, Julio Ma, Yu Tina Zhao, Shougang Zhuang, Paul Y Liu, Lei Wei, Gangjian Qin, Yaoliang Tang, Ting C Zhao
ABSTRACT
We have recently shown that in vivo inhibition of histone deacetylase (HDAC) stimulates endogenous myocardial regeneration in infarcted hearts (Zhang L et al. J Pharmacol Exp Ther 341: 285-293, 2012). Furthermore, our observation demonstrates that HDAC inhibition promotes cardiogenesis, which is associated with HDAC4 reduction. However, it remains unknown as to whether specific inhibition of HDAC4 modulates cardiac stem cells (CSCs) to facilitate myocardial repair and to preserve cardiac performance. c-kit(+) CSCs were isolated from adult mouse hearts and were transfected with HDAC4 siRNA to knockdown HDAC4 of c-kit(+) CSCs. The transfection of HDAC4 siRNA caused a marked reduction of HDAC4 mRNA and proteins in c-kit(+) CSCs. Mouse myocardial infarction (MI) was created to assess the effect of HDAC4 inhibition in c-kit(+) CSCs on myocardial regeneration in vivo when cells were introduced into MI hearts. Transplantation of HDAC4 siRNA-treated c-kit(+) CSCs into MI hearts improved ventricular function, attenuated ventricular remodeling, and promoted CSC-derived regeneration and neovascularization. Furthermore, Ki67 and BrdU positively proliferative myocytes increased in MI hearts receiving HDAC4 siRNA-treated c-kit(+) CSCs compared with MI hearts engrafted with control siRNA-treated c-kit(+) CSCs. In addition, compared with MI hearts engrafted with control adenoviral GFP-infected c-kit(+) CSCs, MI hearts receiving adenoviral HDAC4-infected c-kit(+) CSCs exhibited attenuated cardiac functional recovery, CSC-derived regeneration, and neovascularization, which was accompanied with adverse ventricular remodeling and decrease in Ki67 and BrdU positively proliferative myocytes. HDAC4 inhibition facilitated c-kit(+) CSCs into the differentiation into cardiac lineage commitments in vitro, while HDAC4 overexpression attenuated c-kit(+) CSC-derived cardiogenesis. Our results indicate that HDAC4 inhibition promotes CSC-derived cardiac regeneration and improves the restoration of cardiac function.
Sigma-Aldrich
MISSION® esiRNA, targeting human HDAC4