Merck
  • Somatic mutation of GRIN2A in malignant melanoma results in loss of tumor suppressor activity via aberrant NMDAR complex formation.
Somatic mutation of GRIN2A in malignant melanoma results in loss of tumor suppressor activity via aberrant NMDAR complex formation.
The Journal of investigative dermatology (2014-04-18)
Todd D Prickett, Brad J Zerlanko, Victoria K Hill, Jared J Gartner, Nouar Qutob, Jiji Jiang, May Simaan, John Wunderlich, J Silvio Gutkind, Steven A Rosenberg, Yardena Samuels
ABSTRACT
The ionotropic glutamate receptors (N-methyl-D-aspartate receptors (NMDARs)) are composed of large complexes of multi-protein subunits creating ion channels in the cell plasma membranes that allow for influx or efflux of mono- or divalent cations (e.g., Ca(2+)) important for synaptic transmissions, cellular migration, and survival. Recently, we discovered the high prevalence of somatic mutations within one of the ionotropic glutamate receptors, GRIN2A, in malignant melanoma. Functional characterization of a subset of GRIN2A mutants demonstrated a loss of NMDAR complex formation between GRIN1 and GRIN2A, increased anchorage-independent growth in soft agar, and increased migration. Somatic mutation of GRIN2A results in a dominant negative effect inhibiting the tumor-suppressive phenotype of wild-type (WT) GRIN2A in melanoma. Depletion of endogenous GRIN2A in melanoma cells expressing WT GRIN2A resulted in increased proliferation compared with control. In contrast, short-hairpin RNA depletion of GRIN2A in mutant cell lines slightly reduced proliferation. Our data show that somatic mutation of GRIN2A results in increased survival, and we demonstrate the functional importance of GRIN2A mutations in melanoma and the significance that ionotropic glutamate receptor signaling has in malignant melanoma.
Sigma-Aldrich
Monoclonal Anti-SYP antibody produced in mouse, clone 3B3, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-GAPDH Mouse mAb (6C5), liquid, clone 6C5, Calbiochem®
Pricing and availability is not currently available.