Merck
  • The relation of the dopamine transporter gene (DAT1) to symptoms of internalizing disorders in children.
The relation of the dopamine transporter gene (DAT1) to symptoms of internalizing disorders in children.
Behavior genetics (1998-07-22)
D C Rowe, C Stever, J M Gard, H H Cleveland, M L Sanders, A Abramowitz, S T Kozol, J H Mohr, S L Sherman, I D Waldman
ABSTRACT
The relation of the dopamine transporter gene (DAT1) to symptoms of internalizing disorders, Tourette's disorder, and obsessive-compulsive disorder was examined using both within- and between-family tests of association. The sample consisted of clinic-referred children and their siblings and controls and their siblings. Between-family association was examined via the association of DAT1 genotypes with disorder symptoms in the population. Symptoms of all eight disorders increased with a greater number of 10-repeat DAT1 alleles. Using a quantitative transmission disequilibrium test (QTDT), linkage and within-family association was indicated by increased symptoms in children who received 10 repeat alleles from heterozygous parents relative to children who received 9 repeat alleles. Four disorders were associated with DAT1 using the QTDT: generalized anxiety, social phobia, obsessive-compulsive, and Tourette's. The effects of comorbidity were investigated by repeating the same between- and within-family analyses on residual scores, with any effects of attention deficit hyperactivity disorder symptoms removed. Although the residuals were associated less strongly with DAT1 than were the original scores, three disorders continued to show association both between and within families: generalized anxiety, Tourette's, and social phobia.