Merck
  • Orai3 constitutes a native store-operated calcium entry that regulates non small cell lung adenocarcinoma cell proliferation.
Orai3 constitutes a native store-operated calcium entry that regulates non small cell lung adenocarcinoma cell proliferation.
PloS one (2013-09-24)
Anne-Sophie Ay, Nazim Benzerdjeb, Nazim Benzerdjerb, Henri Sevestre, Ahmed Ahidouch, Halima Ouadid-Ahidouch
ABSTRACT
Orai channels have been associated with cell proliferation, survival and metastasis in several cancers. The present study investigates the expression and the role of Orai3 in cell proliferation in non-small cell lung cancer (NSCLC). We show that Orai3 is over-expressed in cancer tissues as compared to the non-tumoral ones. Furthermore, Orai3 staining is stronger in high grade tumors. Pharmacological inhibition or knockdown of Orai3 significantly reduced store operated calcium entry (SOCE), inhibited cell proliferation and arrested cells of two NSCLC cell lines in G0/G1 phase. These effects were concomitant with a down-regulation of cyclin D1, cyclin E, CDK4 and CDK2 expression. Moreover, Orai3 silencing decreased Akt phosphorylation levels. In conclusion, Orai3 constitutes a native SOCE pathway in NSCLC that controls cell proliferation and cell cycle progression likely via Akt pathway.
Sigma-Aldrich
Anti-ORAI3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.