Merck
  • Reactivity of 3d transition metal cations in diethylene glycol solutions. Synthesis of transition metal ferrites with the structure of discrete nanoparticles complexed with long-chain carboxylate anions.
Reactivity of 3d transition metal cations in diethylene glycol solutions. Synthesis of transition metal ferrites with the structure of discrete nanoparticles complexed with long-chain carboxylate anions.
Inorganic chemistry (2002-11-12)
Daniela Caruntu, Yann Remond, Nam Hawn Chou, Moo-Jin Jun, Gabriel Caruntu, Jibao He, Galina Goloverda, Charles O'Connor, Vladimir Kolesnichenko
ABSTRACT
Study of the reactivity of 3d transition metal cations in diethylene glycol solutions revealed several key features that made it possible to develop a new method for synthesis of the nanocrystalline transition metal ferrites. The 3-7 nm particles of [MFe2O4]n[O2CR]m, where M = Mn, Fe, Co, Ni, and Zn, ligated on their surface with long-chain carboxylate anions, have been obtained in an isolated yield of 75-90%. The key features are the following. Complexation of the first-row transition metal cations with diethylene glycol at a presence of alkaline hydroxide is sufficient to enable control over the rate of their hydrolysis. The reaction of hydrolysis leads to the formation of metal oxide nanocrystals in colloidal solution. The nanoparticles growth is terminated by an added long-chain carboxylic acid, which binds to their surface and acts as a capping ligand. The isolated nanocrystalline powders are stable against agglomeration and highly soluble in nonpolar organic solvents.
Sigma-Aldrich
Diethylene glycol, puriss. p.a., ≥99.0% (GC), colorless
Pricing and availability is not currently available.